Human CD3 epsilon, His Tag (CDE-H5223) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession NP000 724.1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human CD3 epsilon, His Tag (CDE-H5223) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession NP000 724.1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human CD3 epsilon, His Tag (CDE-H5223) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (Accession NP000 724.1). 1 * 100 µG ACRBCDE-H5223100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou