Recombinat Protein,Human E-Selectin CD62E Protein, Fc Tag,Human E-Selectin, Fc Tag (CDE-H5250) is expressed from human 293 cells (H EK293). It contains AA Trp 22 - Phe 556 (Accession NP000441.2).This protein carries a human IgG1 Fc tag at the C-term 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human E-Selectin CD62E Protein, Fc Tag,Human E-Selectin, Fc Tag (CDE-H5250) is expressed from human 293 cells (H EK293). It contains AA Trp 22 - Phe 556 (Accession NP000441.2).This protein carries a human IgG1 Fc tag at the C-term 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human E-Selectin CD62E Protein, Fc Tag,Human E-Selectin, Fc Tag (CDE-H5250) is expressed from human 293 cells (H EK293). It contains AA Trp 22 - Phe 556 (Accession NP000441.2).This protein carries a human IgG1 Fc tag at the C-term 1 * 100 µG ACRBCDE-H5250100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou