Recombinat Protein,Human CD300LG Nepmucin Protein, His Tag,Human CD300LG, His Tag (CDG-H5223) is expressed from human 293 cells (HE K293). It contains AA Leu 19 - Arg 247 (Accession AAH25395).This protein carries a polyhistidine tag at the C-terminu 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD300LG Nepmucin Protein, His Tag,Human CD300LG, His Tag (CDG-H5223) is expressed from human 293 cells (HE K293). It contains AA Leu 19 - Arg 247 (Accession AAH25395).This protein carries a polyhistidine tag at the C-terminu 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD300LG Nepmucin Protein, His Tag,Human CD300LG, His Tag (CDG-H5223) is expressed from human 293 cells (HE K293). It contains AA Leu 19 - Arg 247 (Accession AAH25395).This protein carries a polyhistidine tag at the C-terminu 1 * 100 µG ACRBCDG-H5223100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou