Human CD28H, His Tag (CDH-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession Q96BF3-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human CD28H, His Tag (CDH-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession Q96BF3-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human CD28H, His Tag (CDH-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession Q96BF3-1). 1 * 1 mg ACRBCDH-H52H3-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou