Human CD28H, His Tag (CDH-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession Q96BF3-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human CD28H, His Tag (CDH-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession Q96BF3-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human CD28H, His Tag (CDH-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession Q96BF3-1). 1 * 100 µG ACRBCDH-H52H3100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou