Biotinylated Human LILRB1, Fc,Avitag (CDJ-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - His 458 (Access ion D9IDM8-1). 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human LILRB1, Fc,Avitag (CDJ-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - His 458 (Access ion D9IDM8-1). 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human LILRB1, Fc,Avitag (CDJ-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - His 458 (Access ion D9IDM8-1). 1 * 25 µG ACRBCDJ-H82F7-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou