Recombinat Protein,Mouse LILRB4 CD85k ILT3 Protein, His Tag,Mouse LILRB4, His Tag (CDK-M5229) is expressed from human 293 cells (H EK293). It contains AA Gly 24 - Lys 238 (Accession Q64281-1 ).This protein carries a polyhistidine tag at the C-termin 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse LILRB4 CD85k ILT3 Protein, His Tag,Mouse LILRB4, His Tag (CDK-M5229) is expressed from human 293 cells (H EK293). It contains AA Gly 24 - Lys 238 (Accession Q64281-1 ).This protein carries a polyhistidine tag at the C-termin 1 * 100 µG ACRBCDK-M5229100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou