Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (CDL-M5248) is expressed from human 293 cells (HEK293). It contains AA Gl y 115 - Leu 260 (Accession P27548-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (CDL-M5248) is expressed from human 293 cells (HEK293). It contains AA Gl y 115 - Leu 260 (Accession P27548-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (CDL-M5248) is expressed from human 293 cells (HEK293). It contains AA Gl y 115 - Leu 260 (Accession P27548-1). 1 * 1 mg ACRBCDL-M5248-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou