Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (CDL-M5248) is expressed from human 293 cells (HEK293). It contains AA Gl y 115 - Leu 260 (Accession P27548-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (CDL-M5248) is expressed from human 293 cells (HEK293). It contains AA Gl y 115 - Leu 260 (Accession P27548-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Mouse CD40 Ligand, His Tag (active trimer) (MALS verified) (CDL-M5248) is expressed from human 293 cells (HEK293). It contains AA Gl y 115 - Leu 260 (Accession P27548-1). 1 * 100 µG ACRBCDL-M5248100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou