Human CEACAM-6, His Tag (CE6-H5223) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Gly 320 (Accession NP002474 .3). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human CEACAM-6, His Tag (CE6-H5223) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Gly 320 (Accession NP002474 .3). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human CEACAM-6, His Tag (CE6-H5223) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Gly 320 (Accession NP002474 .3). 1 * 1 mg ACRBCE6-H5223-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou