Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab F(ab)sub2sub. 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab F(ab)sub2sub. 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab F(ab)sub2sub. 1 * 100 µG ACRBCEB-Y27-100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou