Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab F(ab)sub2sub. 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab F(ab)sub2sub. 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with Cetuximab F(ab)sub2sub. 1 * 1 mg ACRBCEB-Y28-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou