Recombinat Protein,Human Complement Factor D CFD Protein, Fc Tag,Human Complement Factor D, Fc Tag (CFD-H5256) is expressed from hu man 293 cells (HEK293) and inactive. It contains AA Ile 26 - Ala 253 (Accession P00746-1).This protein carries a huma 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Complement Factor D CFD Protein, Fc Tag,Human Complement Factor D, Fc Tag (CFD-H5256) is expressed from hu man 293 cells (HEK293) and inactive. It contains AA Ile 26 - Ala 253 (Accession P00746-1).This protein carries a huma 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Complement Factor D CFD Protein, Fc Tag,Human Complement Factor D, Fc Tag (CFD-H5256) is expressed from hu man 293 cells (HEK293) and inactive. It contains AA Ile 26 - Ala 253 (Accession P00746-1).This protein carries a huma 1 * 1 mg ACRBCFD-H5256-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou