Rhesus macaque Cynomolgus Complement Factor D, His Tag (CFD-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 2 6 - Ala 253 (Accession H9EXC1-1). In the region Ile 26 - Ala 253, the AA sequence of Rhesus macaque and Cynomolgus Co 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Rhesus macaque Cynomolgus Complement Factor D, His Tag (CFD-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 2 6 - Ala 253 (Accession H9EXC1-1). In the region Ile 26 - Ala 253, the AA sequence of Rhesus macaque and Cynomolgus Co 1 * 1 mg ACRBCFD-R52H3-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou