Recombinat Protein,Human CG alpha CGA Protein, His Tag,Human CG alpha, His Tag (CGA-H52H2) is expressed from human 293 cells (HEK29 3). It contains AA Ala 25 - Ser 116 (Accession AAH20782).This protein carries a polyhistidine tag at the C-terminus. s 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CG alpha CGA Protein, His Tag,Human CG alpha, His Tag (CGA-H52H2) is expressed from human 293 cells (HEK29 3). It contains AA Ala 25 - Ser 116 (Accession AAH20782).This protein carries a polyhistidine tag at the C-terminus. s 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CG alpha CGA Protein, His Tag,Human CG alpha, His Tag (CGA-H52H2) is expressed from human 293 cells (HEK29 3). It contains AA Ala 25 - Ser 116 (Accession AAH20782).This protein carries a polyhistidine tag at the C-terminus. s 1 * 100 µG ACRBCGA-H52H2100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou