Recombinat Protein,Human Chitinase 3-like 1 CHI3L1 Protein, His Tag,Human CHI3L1, His Tag (CH1-H5228) is expressed from human 293 c ells (HEK293). It contains AA Tyr 22 - Thr 383 (Accession AAH38354).This protein carries a polyhistidine tag at the C 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Chitinase 3-like 1 CHI3L1 Protein, His Tag,Human CHI3L1, His Tag (CH1-H5228) is expressed from human 293 c ells (HEK293). It contains AA Tyr 22 - Thr 383 (Accession AAH38354).This protein carries a polyhistidine tag at the C 1 * 100 µG ACRBCH1-H5228100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou