Recombinat Protein,Human CLEC12A MICL CLL-1 Protein, His Tag,Human CLEC12A, His Tag (CLA-H5245) is expressed from human 293 cells (HEK293). It contains AA His 75 - Ala 275 (Accession NP001193939.1).This protein carries a polyhistidine tag at the N-t 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CLEC12A MICL CLL-1 Protein, His Tag,Human CLEC12A, His Tag (CLA-H5245) is expressed from human 293 cells (HEK293). It contains AA His 75 - Ala 275 (Accession NP001193939.1).This protein carries a polyhistidine tag at the N-t 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CLEC12A MICL CLL-1 Protein, His Tag,Human CLEC12A, His Tag (CLA-H5245) is expressed from human 293 cells (HEK293). It contains AA His 75 - Ala 275 (Accession NP001193939.1).This protein carries a polyhistidine tag at the N-t 1 * 50 µG ACRBCLA-H5245-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou