Human Clusterin, His Tag (CLU-H5227) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Glu 449 (Accession NP00182 2.3). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human Clusterin, His Tag (CLU-H5227) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Glu 449 (Accession NP00182 2.3). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human Clusterin, His Tag (CLU-H5227) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Glu 449 (Accession NP00182 2.3). 1 * 100 µG ACRBCLU-H5227100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou