Recombinat Protein,HCMV Glycoprotein B (gB) Protein, Fc Tag,HCMV Glycoprotein B (gB), Fc Tag (CMB-V5255) Val 23 - Lys 700 (Accession P13201-1) with furin cleavage site mutated from RTKR to TTQT, was produced in human 293 cells (HEK293) at ACROBiosy e 1 * 500 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,HCMV Glycoprotein B (gB) Protein, Fc Tag,HCMV Glycoprotein B (gB), Fc Tag (CMB-V5255) Val 23 - Lys 700 (Accession P13201-1) with furin cleavage site mutated from RTKR to TTQT, was produced in human 293 cells (HEK293) at ACROBiosy e 1 * 500 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,HCMV Glycoprotein B (gB) Protein, Fc Tag,HCMV Glycoprotein B (gB), Fc Tag (CMB-V5255) Val 23 - Lys 700 (Accession P13201-1) with furin cleavage site mutated from RTKR to TTQT, was produced in human 293 cells (HEK293) at ACROBiosy e 1 * 500 µG ACRBCMB-V5255500UG 1 * 500 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou