Recombinat Protein,Human Contactin-2 Tag1 CNTN2 Protein, His Tag,Human Contactin-2, His Tag (CN2-H5226) is expressed from human 29 3 cells (HEK293). It contains AA Ser 31 - Asn 1012 (Accession NP005067.1).This protein carries a polyhistidine tag at s 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Contactin-2 Tag1 CNTN2 Protein, His Tag,Human Contactin-2, His Tag (CN2-H5226) is expressed from human 29 3 cells (HEK293). It contains AA Ser 31 - Asn 1012 (Accession NP005067.1).This protein carries a polyhistidine tag at s 1 * 100 µG ACRBCN2-H5226100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou