Recombinat Protein,Human CNTF R alpha Protein, His Tag,Human CNTF R alpha, His Tag (CNR-H5225) is expressed from human 293 cells (HE K293). It contains AA Gln 23 - Pro 346 (Accession AAH01492).This protein carries a polyhistidine tag at the C-termin 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CNTF R alpha Protein, His Tag,Human CNTF R alpha, His Tag (CNR-H5225) is expressed from human 293 cells (HE K293). It contains AA Gln 23 - Pro 346 (Accession AAH01492).This protein carries a polyhistidine tag at the C-termin 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CNTF R alpha Protein, His Tag,Human CNTF R alpha, His Tag (CNR-H5225) is expressed from human 293 cells (HE K293). It contains AA Gln 23 - Pro 346 (Accession AAH01492).This protein carries a polyhistidine tag at the C-termin 1 * 100 µG ACRBCNR-H5225100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou