Cynomolgus Complement C5, His Tag (CO5-C52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accessio n XP005580972.1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Cynomolgus Complement C5, His Tag (CO5-C52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accessio n XP005580972.1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Cynomolgus Complement C5, His Tag (CO5-C52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Accessio n XP005580972.1). 1 * 100 µG ACRBCO5-C52HX100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou