Human Complement C5 (R885H), His Tag (CO5-H52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Acces sion P01031-1 (R885H)). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human Complement C5 (R885H), His Tag (CO5-H52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Acces sion P01031-1 (R885H)). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human Complement C5 (R885H), His Tag (CO5-H52Hx) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Cys 1676 (Acces sion P01031-1 (R885H)). 1 * 1 mg ACRBCO5-H52HX-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou