Mouse Complement C5, His Tag (CO5-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 1680 (Accession P0 6684-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Mouse Complement C5, His Tag (CO5-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 1680 (Accession P0 6684-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Mouse Complement C5, His Tag (CO5-M52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 1680 (Accession P0 6684-1). 1 * 100 µG ACRBCO5-M52H4100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou