Recombinat Protein,Rabbit Complement C5 Protein, His Tag,Rabbit Complement C5, His Tag (CO5-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Ala 1678 (Accession XP008271599.1).This protein carries a polyhistidine tag at th h 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Rabbit Complement C5 Protein, His Tag,Rabbit Complement C5, His Tag (CO5-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Ala 1678 (Accession XP008271599.1).This protein carries a polyhistidine tag at th h 1 * 1 mg ACRBCO5-R52H4-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou