Recombinat Protein,Human COMP TSP5 Protein, His Tag,Human COMP, His Tag (COP-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Ala 757 (Accession AAI25093).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human COMP TSP5 Protein, His Tag,Human COMP, His Tag (COP-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Ala 757 (Accession AAI25093).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human COMP TSP5 Protein, His Tag,Human COMP, His Tag (COP-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Ala 757 (Accession AAI25093).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 1 mg ACRBCOP-H5220-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou