Recombinat Protein,Human C-Reactive Protein (CRP) Protein, His Tag,Human C-Reactive Protein, His Tag (CRP-H5226) is expressed from h uman 293 cells (HEK293). It contains AA Gln 19 - Pro 224 (Accession NP000558.2).This protein carries a polyhistidine 1 * 250 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human C-Reactive Protein (CRP) Protein, His Tag,Human C-Reactive Protein, His Tag (CRP-H5226) is expressed from h uman 293 cells (HEK293). It contains AA Gln 19 - Pro 224 (Accession NP000558.2).This protein carries a polyhistidine 1 * 250 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human C-Reactive Protein (CRP) Protein, His Tag,Human C-Reactive Protein, His Tag (CRP-H5226) is expressed from h uman 293 cells (HEK293). It contains AA Gln 19 - Pro 224 (Accession NP000558.2).This protein carries a polyhistidine 1 * 250 µG ACRBCRP-H5226250UG 1 * 250 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou