Recombinat Protein,Human Cystatin SA CST2 Protein, His Tag,Human Cystatin SA, His Tag (CS2-H5224) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Ala 141 (Accession AAH62679).This protein carries a polyhistidine tag at the C-ter t 1 * 50 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Cystatin SA CST2 Protein, His Tag,Human Cystatin SA, His Tag (CS2-H5224) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Ala 141 (Accession AAH62679).This protein carries a polyhistidine tag at the C-ter t 1 * 50 µG ACRBCS2-H5224-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou