Human M-CSF R, His Tag (CSR-H5228) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession NP005202. 2). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human M-CSF R, His Tag (CSR-H5228) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession NP005202. 2). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human M-CSF R, His Tag (CSR-H5228) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession NP005202. 2). 1 * 1 mg ACRBCSR-H5228-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou