Recombinat Protein,Rabbit M-CSF R CSF1R CD115 Protein, His Tag,Rabbit M-CSF R, His Tag (CSR-R82H5) is expressed from human 293 cel ls (HEK293). It contains AA Val 20 - Ser 503 (Accession G1TTE0-1).This protein carries a polyhistidine tag at the C-te 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Rabbit M-CSF R CSF1R CD115 Protein, His Tag,Rabbit M-CSF R, His Tag (CSR-R82H5) is expressed from human 293 cel ls (HEK293). It contains AA Val 20 - Ser 503 (Accession G1TTE0-1).This protein carries a polyhistidine tag at the C-te 1 * 100 µG ACRBCSR-R82H5100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou