Biotinylated Cynomolgus Rhesus macaque CTLA-4, Avitag,His Tag (CT4-C82E5) is expressed from human 293 cells (HEK293). It contains A A Ala 37 - Ser 160 (Accession G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus mac 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Cynomolgus Rhesus macaque CTLA-4, Avitag,His Tag (CT4-C82E5) is expressed from human 293 cells (HEK293). It contains A A Ala 37 - Ser 160 (Accession G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus mac 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Cynomolgus Rhesus macaque CTLA-4, Avitag,His Tag (CT4-C82E5) is expressed from human 293 cells (HEK293). It contains A A Ala 37 - Ser 160 (Accession G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus mac 1 * 25 µG ACRBCT4-C82E5-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou