Recombinat Protein,Human Cathepsin B CTSB Protein, His Tag,Human Cathepsin B, His Tag (CTB-H5222) is expressed from human 293 cells (HEK293). It contains AA Arg 18 - Ile 339 (Accession NP001899).Human Cathepsin B, His Tag is fused with a polyhistid i 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Cathepsin B CTSB Protein, His Tag,Human Cathepsin B, His Tag (CTB-H5222) is expressed from human 293 cells (HEK293). It contains AA Arg 18 - Ile 339 (Accession NP001899).Human Cathepsin B, His Tag is fused with a polyhistid i 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Cathepsin B CTSB Protein, His Tag,Human Cathepsin B, His Tag (CTB-H5222) is expressed from human 293 cells (HEK293). It contains AA Arg 18 - Ile 339 (Accession NP001899).Human Cathepsin B, His Tag is fused with a polyhistid i 1 * 1 mg ACRBCTB-H5222-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou