Biotinylated Human CTGF, His,Avitag (recommended for biopanning) (CTF-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession Q5M8T4-1). 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human CTGF, His,Avitag (recommended for biopanning) (CTF-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession Q5M8T4-1). 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human CTGF, His,Avitag (recommended for biopanning) (CTF-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 27 - Ala 349 (Accession Q5M8T4-1). 1 * 25 µG ACRBCTF-H82E6-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou