Mouse CTGF, His Tag (CTF-M52Hc) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Ala 348 (Accession P29268-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Mouse CTGF, His Tag (CTF-M52Hc) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Ala 348 (Accession P29268-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Mouse CTGF, His Tag (CTF-M52Hc) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Ala 348 (Accession P29268-1). 1 * 100 µG ACRBCTF-M52HC100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou