Biotinylated Human CX3CL1, His Tag, primary amine labeling (CX1-H8221) is expressed from human 293 cells (HEK293). It contains AA Gl n 25 - Gln 341 (Accession AAH01163). It is the biotinylated form of Human CX3CL1, His Tag (Cat. No.CX1-H5221). 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human CX3CL1, His Tag, primary amine labeling (CX1-H8221) is expressed from human 293 cells (HEK293). It contains AA Gl n 25 - Gln 341 (Accession AAH01163). It is the biotinylated form of Human CX3CL1, His Tag (Cat. No.CX1-H5221). 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human CX3CL1, His Tag, primary amine labeling (CX1-H8221) is expressed from human 293 cells (HEK293). It contains AA Gl n 25 - Gln 341 (Accession AAH01163). It is the biotinylated form of Human CX3CL1, His Tag (Cat. No.CX1-H5221). 1 * 25 µG ACRBCX1-H8221-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou