Recombinat Protein,Human CXADR CAR Protein, His Tag,Human CXADR, His Tag (CXR-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 237 (Accession NP001329.1).This protein carries a polyhistidine tag at the C-terminus. The ca 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CXADR CAR Protein, His Tag,Human CXADR, His Tag (CXR-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 237 (Accession NP001329.1).This protein carries a polyhistidine tag at the C-terminus. The ca 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CXADR CAR Protein, His Tag,Human CXADR, His Tag (CXR-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 237 (Accession NP001329.1).This protein carries a polyhistidine tag at the C-terminus. The ca 1 * 1 mg ACRBCXR-H5222-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou