Recombinat Protein,Human Decorin Protein, His Tag,Human Decorin, His Tag (DE1-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Lys 359 (Accession NP001911.1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 200 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Decorin Protein, His Tag,Human Decorin, His Tag (DE1-H5223) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Lys 359 (Accession NP001911.1).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 200 µG ACRBDE1-H5223200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou