Recombinat Protein,Human SMAC Diablo Protein, His Tag,Human SMAC, His Tag (DIO-H5129) is expressed from E.coli cells. It contains A A Ala 56 - Asp 239 (Accession NP063940).This protein carries a polyhistidine tag at the C-terminus. The protein has a 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human SMAC Diablo Protein, His Tag,Human SMAC, His Tag (DIO-H5129) is expressed from E.coli cells. It contains A A Ala 56 - Asp 239 (Accession NP063940).This protein carries a polyhistidine tag at the C-terminus. The protein has a 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human SMAC Diablo Protein, His Tag,Human SMAC, His Tag (DIO-H5129) is expressed from E.coli cells. It contains A A Ala 56 - Asp 239 (Accession NP063940).This protein carries a polyhistidine tag at the C-terminus. The protein has a 1 * 200 µG ACRBDIO-H5129200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou