Human Dkk-1, His Tag (DK1-H5221) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accession NP036374.1) . 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human Dkk-1, His Tag (DK1-H5221) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accession NP036374.1) . 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human Dkk-1, His Tag (DK1-H5221) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accession NP036374.1) . 1 * 1 mg ACRBDK1-H5221-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou