Recombinat Protein,Human Dkk-1 Protein, Fc Tag,Human Dkk-1, Fc Tag (DK1-H5258) is expressed from human 293 cells (HEK293). It contai ns AA Thr 32 - His 266 (Accession NP036374.1).This protein carries a human IgG1 Fc tag at the C-terminus. The protei 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Dkk-1 Protein, Fc Tag,Human Dkk-1, Fc Tag (DK1-H5258) is expressed from human 293 cells (HEK293). It contai ns AA Thr 32 - His 266 (Accession NP036374.1).This protein carries a human IgG1 Fc tag at the C-terminus. The protei 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Dkk-1 Protein, Fc Tag,Human Dkk-1, Fc Tag (DK1-H5258) is expressed from human 293 cells (HEK293). It contai ns AA Thr 32 - His 266 (Accession NP036374.1).This protein carries a human IgG1 Fc tag at the C-terminus. The protei 1 * 1 mg ACRBDK1-H5258-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou