Biotinylated Human Dkk-1, Fc,Avitag (DK1-H82F5) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accessi on O94907-1). 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human Dkk-1, Fc,Avitag (DK1-H82F5) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accessi on O94907-1). 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human Dkk-1, Fc,Avitag (DK1-H82F5) is expressed from human 293 cells (HEK293). It contains AA Thr 32 - His 266 (Accessi on O94907-1). 1 * 25 µG ACRBDK1-H82F5-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou