Recombinat Protein,Human DLL1 Delta1 Protein, His Tag,Human DLL1, His Tag (DL1-H52H8) is expressed from human 293 cells (HEK293). I t contains AA Gln 18 - Gly 540 (Accession NP005609).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human DLL1 Delta1 Protein, His Tag,Human DLL1, His Tag (DL1-H52H8) is expressed from human 293 cells (HEK293). I t contains AA Gln 18 - Gly 540 (Accession NP005609).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human DLL1 Delta1 Protein, His Tag,Human DLL1, His Tag (DL1-H52H8) is expressed from human 293 cells (HEK293). I t contains AA Gln 18 - Gly 540 (Accession NP005609).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 100 µG ACRBDL1-H52H8100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou