Recombinat Protein,Human DLL3 Protein, His Tag,Human DLL3, His Tag (DL3-H52H4) is expressed from human 293 cells (HEK293). It contai ns AA Ala 27 - Leu 492 (Accession Q9NYJ7-1).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human DLL3 Protein, His Tag,Human DLL3, His Tag (DL3-H52H4) is expressed from human 293 cells (HEK293). It contai ns AA Ala 27 - Leu 492 (Accession Q9NYJ7-1).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 µG ACRBDL3-H52H4100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou