Recombinat Protein,Human DLL4 Protein, His Tag,Human DLL4, His Tag (DL4-H5227) is expressed from human 293 cells (HEK293). It contai ns AA Ser 27 - Pro 524 (Accession AAI06951).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human DLL4 Protein, His Tag,Human DLL4, His Tag (DL4-H5227) is expressed from human 293 cells (HEK293). It contai ns AA Ser 27 - Pro 524 (Accession AAI06951).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human DLL4 Protein, His Tag,Human DLL4, His Tag (DL4-H5227) is expressed from human 293 cells (HEK293). It contai ns AA Ser 27 - Pro 524 (Accession AAI06951).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 1 mg ACRBDL4-H5227-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou