Human DNAM-1, His Tag (DN1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Accession NP006557). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human DNAM-1, His Tag (DN1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Accession NP006557). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human DNAM-1, His Tag (DN1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Accession NP006557). 1 * 1 mg ACRBDN1-H52H6-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou