Recombinat Protein,Human DPPIV CD26 Protein, Tag Free,Human DPPIV, Tag Free (DP4-H5211) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries no tag. The protein has a calculated MW he 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human DPPIV CD26 Protein, Tag Free,Human DPPIV, Tag Free (DP4-H5211) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries no tag. The protein has a calculated MW he 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human DPPIV CD26 Protein, Tag Free,Human DPPIV, Tag Free (DP4-H5211) is expressed from human 293 cells (HEK293). It contains AA Asn 29 - Pro 766 (Accession NP001926.2).This protein carries no tag. The protein has a calculated MW he 1 * 1 mg ACRBDP4-H5211-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou