Recombinat Protein,Human DPPIV CD26 Protein, His Tag,Human DPPIV, His Tag (DP4-H5221) is expressed from human 293 cells (HEK293). I t contains AA Asp 34 - Pro 766 (Accession NP001926.2).This protein carries a polyhistidine tag at the C-terminus. The 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human DPPIV CD26 Protein, His Tag,Human DPPIV, His Tag (DP4-H5221) is expressed from human 293 cells (HEK293). I t contains AA Asp 34 - Pro 766 (Accession NP001926.2).This protein carries a polyhistidine tag at the C-terminus. The 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human DPPIV CD26 Protein, His Tag,Human DPPIV, His Tag (DP4-H5221) is expressed from human 293 cells (HEK293). I t contains AA Asp 34 - Pro 766 (Accession NP001926.2).This protein carries a polyhistidine tag at the C-terminus. The 1 * 200 µG ACRBDP4-H5221200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou