Recombinat Protein,Human Ephrin-A3 EFNA3 Protein, Fc Tag,Human Ephrin-A3, Fc Tag (EA3-H5258) is expressed from human 293 cells (HEK 293). It contains AA Gln 23 - Ser 213 (Accession AAI10407).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Ephrin-A3 EFNA3 Protein, Fc Tag,Human Ephrin-A3, Fc Tag (EA3-H5258) is expressed from human 293 cells (HEK 293). It contains AA Gln 23 - Ser 213 (Accession AAI10407).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Ephrin-A3 EFNA3 Protein, Fc Tag,Human Ephrin-A3, Fc Tag (EA3-H5258) is expressed from human 293 cells (HEK 293). It contains AA Gln 23 - Ser 213 (Accession AAI10407).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 1 mg ACRBEA3-H5258-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou