Recombinat Protein,Human Ephrin-B1 EFNB1 Protein, His Tag,Human Ephrin-B1, His Tag (EF1-H5223) is expressed from human 293 cells (H EK293). It contains AA Leu 28 - Gly 232 (Accession NP004420).This protein carries a polyhistidine tag at the C-termin 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Ephrin-B1 EFNB1 Protein, His Tag,Human Ephrin-B1, His Tag (EF1-H5223) is expressed from human 293 cells (H EK293). It contains AA Leu 28 - Gly 232 (Accession NP004420).This protein carries a polyhistidine tag at the C-termin 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Ephrin-B1 EFNB1 Protein, His Tag,Human Ephrin-B1, His Tag (EF1-H5223) is expressed from human 293 cells (H EK293). It contains AA Leu 28 - Gly 232 (Accession NP004420).This protein carries a polyhistidine tag at the C-termin 1 * 200 µG ACRBEF1-H5223200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou