Recombinat Protein,Human Ephrin-A1 EFNA1 Protein, Fc Tag,Human Ephrin-A1, Fc Tag (EF1-H5251) is expressed from human 293 cells (HEK 293). It contains AA Asp 19 - Ser 182 (Accession NP004419).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Ephrin-A1 EFNA1 Protein, Fc Tag,Human Ephrin-A1, Fc Tag (EF1-H5251) is expressed from human 293 cells (HEK 293). It contains AA Asp 19 - Ser 182 (Accession NP004419).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Ephrin-A1 EFNA1 Protein, Fc Tag,Human Ephrin-A1, Fc Tag (EF1-H5251) is expressed from human 293 cells (HEK 293). It contains AA Asp 19 - Ser 182 (Accession NP004419).This protein carries a human IgG1 Fc tag at the C-terminus 1 * 1 mg ACRBEF1-H5251-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou